Gene Transcripts

Gene Symbol
EPS8
Entrez Gene ID
2059
Gene Name
epidermal growth factor receptor pathway substrate 8
Gene Aliases
DFNB102
Chromosome Location
Chr. 12: 15620141 - 15789576 on Build GRCh38
UniGene ID
Hs.591160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004447.5 NP_004438.3 5 778
XM_011520605.2 XP_011518907.1 6 838
GenBank mRNA AK291777.1 - 5 583
AK292931.1 - 5 435
AK304860.1 - 5 778
BC030010.2 - 5 590 4820933
U12535.1 - 5 550

View Full Product Details