Gene Transcripts
- Gene Symbol
- FANCC
- Entrez Gene ID
- 2176
- Gene Name
- Fanconi anemia complementation group C
- Gene Aliases
- FA3, FAC, FACC
- Chromosome Location
- Chr. 9: 95099054 - 95317730 on Build GRCh38
- UniGene ID
- Hs.494529
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000136.2 |
NP_000127.2
|
4, 5 |
610 |
|
NM_001243743.1 |
NP_001230672.1
|
4, 5 |
552 |
|
NM_001243744.1 |
NP_001230673.1
|
4, 5 |
610 |
|
XM_006717001.2 |
XP_006717064.1
|
4, 5 |
592 |
|
XM_006717002.3 |
XP_006717065.1
|
4, 5 |
592 |
|
XM_006717004.3 |
XP_006717067.1
|
4, 5 |
592 |
|
XM_011518365.2 |
XP_011516667.1
|
4, 5 |
585 |
|
XM_011518366.2 |
XP_011516668.1
|
4, 5 |
592 |
|
GenBank mRNA |
AK222871.1 |
- |
4, 5 |
572 |
|
AK312548.1 |
- |
4, 5 |
476 |
|
AU132608.1 |
- |
4, 5 |
610 |
|
BC015748.1 |
- |
4, 5 |
600 |
4864916 |
KU178051.1 |
- |
3, 4 |
348 |
|
X66893.1 |
- |
4, 5 |
603 |
|
X66894.1 |
- |
4, 5 |
521 |
|
View Full Product Details