Gene Transcripts

Gene Symbol
STT3A
Entrez Gene ID
3703
Gene Name
STT3A, catalytic subunit of the oligosaccharyltransferase complex
Gene Aliases
ITM1, STT3-A, TMC
Chromosome Location
Chr. 11: 125592795 - 125622759 on Build GRCh38
UniGene ID
Hs.504237
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278503.1 NP_001265432.1 11 1317
NM_001278504.1 NP_001265433.1 9 1106
NM_152713.4 NP_689926.1 10 1229
XM_011542807.2 XP_011541109.1 10 1256
GenBank mRNA AK290040.1 - 10 1179
AK290657.1 - 10 1177
AK295892.1 - 9 1034
AK308902.1 - 10 1172
BC020965.2 - 10 1162 3891543
BC048348.1 - 9 1115 5561615
BT007100.1 - 9 1070
BX649102.1 - 11 1255
L38961.1 - 10 1176
L47337.1 - 10 1186

View Full Product Details