Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000286.2 | NP_000277.1 | 2, 3 | 1299 | |
GenBank mRNA | AB004546.1 | - | 2, 3 | 868 | |
AK312635.1 | - | 2, 3 | 1212 | ||
BC015751.1 | - | 2, 3 | 1214 | ||
BC031085.1 | - | 2, 3 | 1203 | 5296541 | |
U91521.1 | - | 2, 3 | 1230 |