Gene Transcripts

Gene Symbol
PRSS8
Entrez Gene ID
5652
Gene Name
protease, serine 8
Gene Aliases
CAP1, PROSTASIN
Chromosome Location
Chr. 16: 31131433 - 31135762 on Build GRCh38
UniGene ID
Hs.75799
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002773.4 NP_002764.1 5 854
GenBank mRNA AK296656.1 - 5 573
AK301619.1 - 5 657
BC001462.2 - 5 765 3138532
L41351.1 - 5 818

View Full Product Details