Gene Transcripts
- Gene Symbol
- STIM1
- Entrez Gene ID
- 6786
- Gene Name
- stromal interaction molecule 1
- Gene Aliases
- D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
- Chromosome Location
- Chr. 11: 3855703 - 4093210 on Build GRCh38
- UniGene ID
- Hs.501735
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U52426.1 |
- |
12 |
2581 |
|
View Full Product Details