Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007144.2 | NP_009075.1 | 3, 4 | 349 | |
XM_005257640.2 | XP_005257697.1 | 4, 5 | 808 | ||
XM_005257641.4 | XP_005257698.1 | 2, 3 | 1030 | ||
XM_005257642.3 | XP_005257699.1 | 2, 3 | 1427 | ||
XM_017025016.1 | XP_016880505.1 | 4, 5 | 1368 | ||
GenBank mRNA | BC004858.2 | - | 1, 2 | 120 | |
BC024255.1 | - | 3, 4 | 345 | 3605568 | |
BX647429.1 | - | 4, 5 | 808 | ||
D13969.1 | - | 2, 3 | 307 |