Gene Transcripts
- Gene Symbol
- FAM13A
- Entrez Gene ID
- 10144
- Gene Name
- family with sequence similarity 13 member A
- Gene Aliases
- ARHGAP48, FAM13A1
- Chromosome Location
- Chr. 4: 88725954 - 89111398 on Build GRCh38
- UniGene ID
- Hs.714536
- Species
- Homo sapiens
View Full Product Details