Gene Transcripts
- Gene Symbol
- COPS5
- Entrez Gene ID
- 10987
- Gene Name
- COP9 signalosome subunit 5
- Gene Aliases
- CSN5, JAB1, MOV-34, SGN5
- Chromosome Location
- Chr. 8: 67043079 - 67062327 on Build GRCh38
- UniGene ID
- Hs.491912
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006837.2 |
NP_006828.2
|
3, 4 |
845 |
|
GenBank mRNA |
AB209134.1 |
- |
2, 3 |
2555 |
|
AK056967.1 |
- |
5, 6 |
621 |
|
AK128519.1 |
- |
2, 3 |
2837 |
|
AK310434.1 |
- |
3, 4 |
647 |
|
BC001187.1 |
- |
3, 4 |
649 |
3354130 |
BC001859.2 |
- |
3, 4 |
627 |
3530890 |
BC007272.1 |
- |
3, 4 |
623 |
3161025 |
BX648542.1 |
- |
3, 4 |
3182 |
|
CR541678.1 |
- |
3, 4 |
514 |
|
U65928.1 |
- |
3, 4 |
641 |
|
U70734.1 |
- |
3, 4 |
634 |
|
View Full Product Details