Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300779.1 | NP_001287708.1 | 5 | 670 | |
NM_015954.3 | NP_057038.2 | 5 | 670 | ||
GenBank mRNA | AF132960.1 | - | 5 | 531 | |
AK024614.1 | - | 5 | 486 | ||
AK222541.1 | - | 5 | 486 | ||
AK296697.1 | - | 5 | 427 | ||
AK309182.1 | - | 5 | 586 | ||
BC041328.1 | - | 5 | 670 | ||
BC056234.1 | - | 5 | 512 | ||
BU928832.1 | - | 5 | 476 |