Gene Transcripts

Gene Symbol
WDR83OS
Entrez Gene ID
51398
Gene Name
WD repeat domain 83 opposite strand
Gene Aliases
C19orf56, PTD008
Chromosome Location
Chr. 19: 12668067 - 12671023 on Build GRCh38
UniGene ID
Hs.108969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016145.3 NP_057229.1 2 389
XM_017026864.1 XP_016882353.1 3 1383
GenBank mRNA AF059620.1 - 2 157
AF078861.1 - 2 374
AF151898.1 - 2 373
AK091030.1 - 2 2094
AK311944.1 - 2 153
BC001192.1 - 2 137
BC042919.1 - 2 158 5932109
BC061912.1 - 2 158 4444565

View Full Product Details