Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018178.5 | NP_060648.2 | 5 | 856 | |
XM_006711428.1 | XP_006711491.1 | 5 | 783 | ||
GenBank mRNA | AJ296153.1 | - | 5 | 684 | |
AK001549.1 | - | 4 | 484 | ||
AK300635.1 | - | 4 | 682 | ||
AK315594.1 | - | 5 | 823 | ||
BC013870.2 | - | 4 | 655 | 3837466 |