Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166412.1 | NP_001159884.1 | 2 | 339 | |
NM_022138.2 | NP_071421.1 | 2 | 339 | ||
XM_011536065.1 | XP_011534367.1 | 2 | 337 | ||
XM_011536066.1 | XP_011534368.1 | 2 | 337 | ||
GenBank mRNA | AB014730.1 | - | 2 | 139 | |
AB014737.1 | - | 2 | 139 | ||
AJ420521.1 | - | 2 | 319 | ||
AK056700.1 | - | 2 | 266 | ||
AK314613.1 | - | 2 | 306 | ||
AL832303.1 | - | 2 | 241 | ||
BC028420.1 | - | 2 | 342 | 4826774 | |
BC047583.1 | - | 2 | 311 | 4826950 |