Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166412.1 | NP_001159884.1 | 3 | 505 | |
NM_022138.2 | NP_071421.1 | 3 | 505 | ||
XM_011536065.1 | XP_011534367.1 | 3 | 503 | ||
XM_011536066.1 | XP_011534368.1 | 3 | 503 | ||
GenBank mRNA | AB014730.1 | - | 3 | 305 | |
AB014737.1 | - | 3 | 305 | ||
AF173892.1 | - | 2 | 160 | ||
AJ420521.1 | - | 3 | 485 | ||
AK056700.1 | - | 3 | 432 | ||
AK314613.1 | - | 3 | 472 | ||
AL832303.1 | - | 3 | 407 | ||
BC028420.1 | - | 3 | 508 | 4826774 | |
BC047583.1 | - | 3 | 477 | 4826950 |