Gene Transcripts
- Gene Symbol
- ZYG11B
- Entrez Gene ID
- 79699
- Gene Name
- zyg-11 family member B, cell cycle regulator
- Gene Aliases
- ZYG11
- Chromosome Location
- Chr. 1: 52726454 - 52827342 on Build GRCh38
- UniGene ID
- Hs.476280
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024646.2 |
NP_078922.1
|
11, 12 |
2086 |
|
XM_006710898.3 |
XP_006710961.1
|
11, 12 |
1973 |
|
GenBank mRNA |
AB051517.1 |
- |
11, 12 |
2036 |
|
AK023518.1 |
- |
5, 6 |
543 |
|
AK056645.1 |
- |
9, 10 |
1030 |
|
AK095643.1 |
- |
10, 11 |
1726 |
|
AK296460.1 |
- |
9, 10 |
1881 |
|
BC029832.1 |
- |
9, 10 |
1100 |
|
BX647643.1 |
- |
9, 10 |
1259 |
|
DQ646393.1 |
- |
12, 13 |
2159 |
|
View Full Product Details