Gene Transcripts
- Gene Symbol
- SNX27
- Entrez Gene ID
- 81609
- Gene Name
- sorting nexin family member 27
- Gene Aliases
- MRT1, MY014
- Chromosome Location
- Chr. 1: 151612027 - 151699083 on Build GRCh38
- UniGene ID
- Hs.192326
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030918.5 |
NP_112180.4
|
6, 7 |
1006 |
|
XM_005245509.2 |
XP_005245566.1
|
6, 7 |
1128 |
|
XM_005245510.3 |
XP_005245567.1
|
6, 7 |
738 |
|
XM_005245511.4 |
XP_005245568.1
|
6, 7 |
1034 |
|
XM_011510024.2 |
XP_011508326.1
|
5, 6 |
849 |
|
XM_011510025.2 |
XP_011508327.1
|
5, 6 |
707 |
|
XM_011510026.2 |
XP_011508328.1
|
6, 7 |
1102 |
|
XM_017002417.1 |
XP_016857906.1
|
5, 6 |
707 |
|
GenBank mRNA |
AB007957.2 |
- |
6, 7 |
975 |
|
AY044866.1 |
- |
5, 6 |
711 |
|
BC012184.1 |
- |
6, 7 |
996 |
|
BC051817.1 |
- |
3, 4 |
255 |
|
BC071825.1 |
- |
5, 6 |
469 |
|
BC100998.1 |
- |
6, 7 |
1006 |
40006277 |
BC100999.1 |
- |
6, 7 |
1004 |
40006279 |
BC101000.1 |
- |
6, 7 |
1006 |
40006281 |
BC101822.1 |
- |
6, 7 |
1041 |
8069328 |
BC101824.1 |
- |
6, 7 |
1041 |
8069330 |
BC107862.1 |
- |
4, 5 |
462 |
|
View Full Product Details