Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171971.2 | NP_001165442.1 | 14 | 1648 | |
NM_033100.3 | NP_149091.1 | 14 | 1648 | ||
XM_011540337.1 | XP_011538639.1 | 13 | 2139 | ||
XM_011540338.1 | XP_011538640.1 | 13 | 2139 | ||
XM_011540339.1 | XP_011538641.1 | 14 | 1454 | ||
XM_011540340.2 | XP_011538642.1 | 13 | 2139 | ||
GenBank mRNA | AB053448.1 | - | 14 | 1521 | |
AB593154.1 | - | 14 | 1648 | ||
AB593155.1 | - | 14 | 1634 | ||
AK124752.1 | - | 7 | 737 | ||
BC038799.1 | - | 14 | 1638 | 5492127 |