Gene Transcripts

Gene Symbol
OR7D4
Entrez Gene ID
125958
Gene Name
olfactory receptor family 7 subfamily D member 4
Gene Aliases
OR19-7, OR19-B, OR19B, OR7D4P, hg105
Chromosome Location
Chr. 19: 9213850 - 9214871 on Build GRCh38
UniGene ID
Hs.446910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005191.2 NP_001005191.1 1 620
GenBank mRNA BC137147.1 - 1 620
BC137162.1 - 1 620 9021159

View Full Product Details