Gene Transcripts
- Gene Symbol
- PIFO
- Entrez Gene ID
- 128344
- Gene Name
- primary cilia formation
- Gene Aliases
- C1orf88, pitchfork
- Chromosome Location
- Chr. 1: 111324663 - 111353017 on Build GRCh38
- UniGene ID
- Hs.172510
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300831.1 |
NP_001287760.1
|
1, 2 |
120 |
|
NM_181643.5 |
NP_857594.2
|
1, 2 |
120 |
|
NR_125338.1 |
|
1, 2 |
120 |
|
GenBank mRNA |
AK074433.1 |
- |
1, 2 |
55 |
|
AK303897.1 |
- |
1, 2 |
81 |
|
AK310801.1 |
- |
1, 2 |
71 |
|
BC050319.1 |
- |
1, 2 |
84 |
4837146 |
BC068603.1 |
- |
1, 2 |
44 |
|
BC093891.1 |
- |
1, 2 |
107 |
7939736 |
BC101501.1 |
- |
1, 2 |
108 |
8069007 |
BF674731.1 |
- |
1, 2 |
121 |
|
HM237139.1 |
- |
1, 2 |
107 |
|
HM237140.1 |
- |
1, 2 |
33 |
|
View Full Product Details