Gene Transcripts
- Gene Symbol
- C2orf73
- Entrez Gene ID
- 129852
- Gene Name
- chromosome 2 open reading frame 73
- Gene Aliases
- Chromosome Location
- Chr. 2: 54329961 - 54383673 on Build GRCh38
- UniGene ID
- Hs.732626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100396.1 |
NP_001093866.1
|
4, 5 |
546 |
|
XM_005264133.1 |
XP_005264190.1
|
3, 4 |
338 |
|
XM_005264134.3 |
XP_005264191.1
|
3, 4 |
421 |
|
XM_011532510.2 |
XP_011530812.1
|
4, 5 |
616 |
|
XM_011532513.2 |
XP_011530815.1
|
3, 4 |
519 |
|
XM_017003323.1 |
XP_016858812.1
|
4, 5 |
441 |
|
GenBank mRNA |
AK097617.1 |
- |
3, 4 |
338 |
|
BC031669.1 |
- |
4, 5 |
509 |
|
BC126244.1 |
- |
3, 4 |
307 |
8991960 |
BC126246.1 |
- |
3, 4 |
307 |
8991962 |
BC144192.1 |
- |
4, 5 |
341 |
9052712 |
View Full Product Details