Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133264.4 | NP_573571.1 | 7 | 1469 | |
XM_005257083.2 | XP_005257140.1 | 7 | 1471 | ||
XM_005257084.2 | XP_005257141.1 | 7 | 1429 | ||
XM_011524412.1 | XP_011522714.1 | 6 | 1370 | ||
XM_011524413.1 | XP_011522715.1 | 6 | 1351 | ||
GenBank mRNA | AB043786.1 | - | 6 | 1229 | |
AF370396.1 | - | 7 | 1394 | ||
AJ431177.1 | - | 6 | 1229 | ||
AK026913.1 | - | 5 | 917 | ||
AK055057.1 | - | 5 | 686 | ||
AK289578.1 | - | 7 | 1429 | ||
AK289779.1 | - | 7 | 1488 | ||
AK290301.1 | - | 7 | 1465 | ||
AK315836.1 | - | 7 | 1449 | ||
AL834239.1 | - | 6 | 1338 | ||
BC025965.1 | - | 7 | 1425 | ||
BC065551.1 | - | 7 | 1406 | 6044222 |