Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321298.1 | NP_001308227.1 | 4 | 678 | |
NM_173637.3 | NP_775908.2 | 3 | 580 | ||
NR_135612.1 | 3 | 580 | |||
XM_011527926.1 | XP_011526228.1 | 3 | 374 | ||
GenBank mRNA | AK097761.1 | - | 4 | 662 | |
BC031671.1 | - | 3 | 580 | 5164630 |