Gene Transcripts

Gene Symbol
OR5F1
Entrez Gene ID
338674
Gene Name
olfactory receptor family 5 subfamily F member 1
Gene Aliases
OR11-10
Chromosome Location
Chr. 11: 55993681 - 55994625 on Build GRCh38
UniGene ID
Hs.553702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003697.1 NP_003688.1 1 62
GenBank mRNA BC101239.2 - 1 64
BC101241.1 - 1 62
BC101242.3 - 1 61

View Full Product Details