Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130009.2 | NP_001123481.2 | 14 | 2345 | |
NM_182625.4 | NP_872431.4 | 14 | 2269 | ||
XM_005262613.4 | XP_005262670.1 | 14 | 2355 | ||
XM_006712005.3 | XP_006712068.1 | 14 | 2252 | ||
XM_011532820.2 | XP_011531122.1 | 14 | 2272 | ||
XM_011532821.2 | XP_011531123.1 | 14 | 2362 | ||
XM_011532822.2 | XP_011531124.1 | 14 | 2242 | ||
GenBank mRNA | AK098188.1 | - | 7 | 1295 | |
AK131387.1 | - | 14 | 2217 | ||
BC117204.1 | - | 13 | 2167 | 40125755 | |
BC117206.1 | - | 13 | 2167 | 40125757 | |
CR936783.1 | - | 14 | 2355 |