Gene Transcripts

Gene Symbol
KRTAP26-1
Entrez Gene ID
388818
Gene Name
keratin associated protein 26-1
Gene Aliases
Chromosome Location
Chr. 21: 30319131 - 30320289 on Build GRCh38
UniGene ID
Hs.28704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203405.1 NP_981950.1 1 484
GenBank mRNA AB096936.1 - 1 245
AM941740.1 - 1 267
BC057825.1 - 1 485 30331793
CB996254.1 - 538

View Full Product Details