Gene Transcripts
- Gene Symbol
- COL9A2
- Entrez Gene ID
- 1298
- Gene Name
- collagen type IX alpha 2
- Gene Aliases
- DJ39G22.4, EDM2, MED, STL5
- Chromosome Location
- Chr. 1: 40300487 - 40317653 on Build GRCh38
- UniGene ID
- Hs.418012
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001852.3 |
NP_001843.1
|
4, 5 |
312 |
|
XM_006710365.3 |
XP_006710428.1
|
5, 6 |
420 |
|
XM_011540715.2 |
XP_011539017.1
|
4, 5 |
888 |
|
XM_011540716.2 |
XP_011539018.1
|
3, 4 |
368 |
|
XM_011540717.2 |
XP_011539019.1
|
4, 5 |
885 |
|
XM_017000332.1 |
XP_016855821.1
|
4, 5 |
354 |
|
XM_017000333.1 |
XP_016855822.1
|
3, 4 |
366 |
|
GenBank mRNA |
BC041479.1 |
- |
4, 5 |
202 |
|
BC136326.1 |
- |
4, 5 |
305 |
9020313 |
BC136327.1 |
- |
4, 5 |
305 |
9020314 |
M95610.1 |
- |
4, 5 |
201 |
|
View Full Product Details