Gene Transcripts

Gene Symbol
FTH1
Entrez Gene ID
2495
Gene Name
ferritin heavy chain 1
Gene Aliases
FHC, FTH, FTHL6, HFE5, PIG15, PLIF
Chromosome Location
Chr. 11: 61964285 - 61967660 on Build GRCh38
UniGene ID
Hs.524910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002032.2 NP_002023.2 4 904
GenBank mRNA AB062402.1 - 4 857
AY258285.1 - 1462
BC000857.1 - 4 863 3459353
BC001399.2 - 4 832 3051458
BC011359.2 - 4 854 4157296
BC013724.2 - 4 857 3857790
BC015156.1 - 4 727 3883694
BC016009.1 - 4 879 4666545
BC016857.1 - 4 823 3848510
BC020300.1 - 4 840
BC035441.1 - 4 863
BC063514.1 - 4 846 3882345
BC066341.1 - 5 781 5288688
BC066961.1 - 4 877 4796312
BC070494.1 - 5 818
BC073750.1 - 4 861 6256153
BC104643.1 - 4 879 4043391
BC105802.1 - 5 856
BM971420.1 - 1 338
L20941.1 - 4 877
M11146.1 - 4 746
M12937.1 - 4 693
M97164.1 - 4 798
X00318.1 - 4 757

Gene Symbol
BEST1
Entrez Gene ID
7439
Gene Name
bestrophin 1
Gene Aliases
ARB, BEST, BMD, RP50, TU15B, VMD2
Chromosome Location
Chr. 11: 61946722 - 61964465 on Build GRCh38
UniGene ID
Hs.524910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA CA388926.1 - 1 151

View Full Product Details