Gene Transcripts
- Gene Symbol
- SNAPC2
- Entrez Gene ID
- 6618
- Gene Name
- small nuclear RNA activating complex polypeptide 2
- Gene Aliases
- PTFDELTA, SNAP45
- Chromosome Location
- Chr. 19: 7920309 - 7923251 on Build GRCh38
- UniGene ID
- Hs.631860
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003083.3 |
NP_003074.1
|
3, 4 |
421 |
|
NR_030717.1 |
|
3, 4 |
318 |
|
GenBank mRNA |
AK222926.1 |
- |
3, 4 |
392 |
|
AK314878.1 |
- |
3, 4 |
406 |
|
BC011868.2 |
- |
3, 4 |
376 |
4545178 |
BP234322.1 |
- |
3, 4 |
317 |
|
BX647416.1 |
- |
3, 4 |
285 |
|
CF454397.1 |
- |
3, 4 |
421 |
|
U44755.1 |
- |
3, 4 |
405 |
|
U44898.1 |
- |
3, 4 |
387 |
|
View Full Product Details