Gene Transcripts

Gene Symbol
TPD52L2
Entrez Gene ID
7165
Gene Name
tumor protein D52 like 2
Gene Aliases
D54, TPD54
Chromosome Location
Chr. 20: 63865228 - 63891545 on Build GRCh38
UniGene ID
Hs.473296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243891.1 NP_001230820.1 5 522
NM_001243892.1 NP_001230821.1 4 468
NM_001243895.1 NP_001230824.1 5 597
NM_003288.3 NP_003279.2 5 597
NM_199359.2 NP_955391.1 4 537
NM_199360.2 NP_955392.1 5 597
NM_199361.2 NP_955393.1 4 537
NM_199362.2 NP_955394.1 5 597
NM_199363.2 NP_955395.1 4 537
NR_045090.1 5 566
XM_011529034.2 XP_011527336.1 5 597
XM_011529035.2 XP_011527337.1 5 597
XM_017028038.1 XP_016883527.1 4 522
GenBank mRNA AB062396.1 - 1 55
AF004430.1 - 5 535
AK055068.1 - 4 496
AK223031.1 - 5 509
AK293349.1 - 5 419
AK297064.1 - 4 495
AK298366.1 - 4 453
AK314501.1 - 5 555
AU124850.1 - 5 526
AY323819.1 - 5 466
BC006804.1 - 5 482 3446037
BI667351.1 - 4 349
BI754201.1 - 4 424
BM473873.1 - 5 554
BP347669.1 - 4 435
BQ948007.1 - 5 492
BT019631.1 - 5 459
CR542036.1 - 5 459
CR749208.1 - 4 436
DA547106.1 - 5 498
DC362382.1 - 4 453
DC428780.1 - 5 452

View Full Product Details