Gene Transcripts

Gene Symbol
BHLHE22
Entrez Gene ID
27319
Gene Name
basic helix-loop-helix family member e22
Gene Aliases
BHLHB5, Beta3, CAGL85, TNRC20
Chromosome Location
Chr. 8: 64580238 - 64583634 on Build GRCh38
UniGene ID
Hs.388788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152414.4 NP_689627.1 1 1684
GenBank mRNA AK091333.1 - 1 759
AK293979.1 - 1 1565

View Full Product Details