Gene Transcripts
- Gene Symbol
- ENY2
- Entrez Gene ID
- 56943
- Gene Name
- ENY2, transcription and export complex 2 subunit
- Gene Aliases
- DC6, Sus1, e(y)2
- Chromosome Location
- Chr. 8: 109334323 - 109345960 on Build GRCh38
- UniGene ID
- Hs.492555
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193557.1 |
NP_001180486.1
|
2, 3 |
168 |
|
NM_020189.5 |
NP_064574.1
|
2, 3 |
240 |
|
NR_036471.1 |
|
2, 3 |
240 |
|
NR_036472.1 |
|
2, 3 |
240 |
|
GenBank mRNA |
AF173296.1 |
- |
2, 3 |
310 |
|
AF201940.1 |
- |
2, 3 |
255 |
|
AK000487.1 |
- |
2, 3 |
144 |
|
AK095651.1 |
- |
2, 3 |
239 |
|
AK290752.1 |
- |
2, 3 |
184 |
|
AK316560.1 |
- |
2, 3 |
235 |
|
BC007870.2 |
- |
2, 3 |
156 |
4303290 |
BX648621.1 |
- |
1 |
989 |
|
CR457183.1 |
- |
1, 2 |
94 |
|
DA548665.1 |
- |
2, 3 |
114 |
|
DA762166.1 |
- |
2, 3 |
222 |
|
DB168034.1 |
- |
2, 3 |
240 |
|
View Full Product Details