Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020319.2 | NP_064715.1 | 9 | 1352 | |
XM_011515458.1 | XP_011513760.1 | 10 | 1479 | ||
GenBank mRNA | AK001389.1 | - | 10 | 1321 | |
AK001740.1 | - | 10 | 1402 | ||
AK023704.1 | - | 9 | 1289 | ||
AL050390.1 | - | 9 | 1294 | ||
BC015453.1 | - | 3 | 270 | ||
BC035353.1 | - | 9 | 1308 | 5185501 |