Gene Transcripts

Gene Symbol
GALP
Entrez Gene ID
85569
Gene Name
galanin like peptide
Gene Aliases
Chromosome Location
Chr. 19: 56176020 - 56185775 on Build GRCh38
UniGene ID
Hs.283915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145546.1 NP_001139018.1 5 500
NM_033106.3 NP_149097.1 6 549
GenBank mRNA AF188492.1 - 6 549
AF188493.1 - 6 549

View Full Product Details