Gene Transcripts

Gene Symbol
ZCWPW2
Entrez Gene ID
152098
Gene Name
zinc finger CW-type and PWWP domain containing 2
Gene Aliases
ZCW2
Chromosome Location
Chr. 3: 28349149 - 28526366 on Build GRCh38
UniGene ID
Hs.510635
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040432.3 NP_001035522.1 8, 9 975
NM_001324169.1 NP_001311098.1 7, 8 855
NR_136708.1 6, 7 737
XM_017005755.1 XP_016861244.1 7, 8 4650
GenBank mRNA BC065764.1 - 6, 7 443
BC094696.1 - 8, 9 955 30529418

View Full Product Details