Gene Transcripts

Gene Symbol
SBDSP1
Entrez Gene ID
155370
Gene Name
Shwachman-Bodian-Diamond syndrome pseudogene 1
Gene Aliases
SBDSP
Chromosome Location
Chr. 7: 72829373 - 72837399 on Build GRCh38
UniGene ID
Hs.585053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001588.2 2, 3 522
NR_024110.1 2, 3 522
GenBank mRNA CX867149.1 - 2, 3 414
DA143511.1 - 2, 3 525

View Full Product Details