Gene Transcripts

Gene Symbol
BTBD17
Entrez Gene ID
388419
Gene Name
BTB domain containing 17
Gene Aliases
BTBD17A, LGALS3BPL, TANGO10A
Chromosome Location
Chr. 17: 74356416 - 74363655 on Build GRCh38
UniGene ID
Hs.211870
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080466.1 NP_001073935.1 3 1067
XM_011524791.2 XP_011523093.1 5 1973
XM_017024622.1 XP_016880111.1 5 1909

View Full Product Details