Gene Transcripts

Gene Symbol
PLA2G2C
Entrez Gene ID
391013
Gene Name
phospholipase A2 group IIC
Gene Aliases
Chromosome Location
Chr. 1: 20163085 - 20177364 on Build GRCh38
UniGene ID
Hs.512512
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316722.1 NP_001303651.1 4 332
XM_017001270.1 XP_016856759.1 2 1001

View Full Product Details