Gene Transcripts

Gene Symbol
TMEM9B
Entrez Gene ID
56674
Gene Name
TMEM9 domain family member B
Gene Aliases
C11orf15
Chromosome Location
Chr. 11: 8947201 - 8965011 on Build GRCh38
UniGene ID
Hs.501853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286094.1 NP_001273023.1 1 98
NM_020644.2 NP_065695.1 1 98
GenBank mRNA BP242422.1 - 1 98
DA563443.1 - 1 65

View Full Product Details