Gene Transcripts
- Gene Symbol
- SEPT7P2
- Entrez Gene ID
- 641977
- Gene Name
- septin 7 pseudogene 2
- Gene Aliases
- SEPT13, SEPT7B
- Chromosome Location
- Chr. 7: 45723787 - 45769018 on Build GRCh38
- UniGene ID
- Hs.723477
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056032.1 |
- |
1 |
802 |
|
View Full Product Details