Gene Transcripts
- Gene Symbol
- KRT17P5
- Entrez Gene ID
- 339240
- Gene Name
- keratin 17 pseudogene 5
- Gene Aliases
- Chromosome Location
- Chr. 17: 18422181 - 18425333 on Build GRCh38
- UniGene ID
- Hs.420616
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001443.1 |
|
1 |
327 |
|
GenBank mRNA |
AK127974.1 |
- |
4 |
1224 |
|
BC040601.1 |
- |
3 |
1228 |
|
- Gene Symbol
- KRT42P
- Entrez Gene ID
- 284116
- Gene Name
- keratin 42 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 17: 41626327 - 41640199 on Build GRCh38
- UniGene ID
- Hs.725790
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095281.1 |
- |
5 |
976 |
|
View Full Product Details