Gene Transcripts
- Gene Symbol
- FEZF1-AS1
- Entrez Gene ID
- 154860
- Gene Name
- FEZF1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 122303658 - 122310077 on Build GRCh38
- UniGene ID
- Hs.664204
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036484.1 |
|
3 |
1295 |
|
GenBank mRNA |
AK057037.1 |
- |
3 |
1295 |
|
View Full Product Details