Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039876.2 | NP_001034965.1 | 8 | 1293 | |
NM_001297735.2 | NP_001284664.1 | 6 | 954 | ||
GenBank mRNA | AA991508.1 | - | 1 | 79 | |
AK093764.1 | - | 7 | 2252 | ||
BC052573.1 | - | 6 | 944 | 6568945 | |
CB241656.1 | - | 1 | 79 |