Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317335.1 | NP_001304264.1 | 2 | 175 | |
NM_001928.3 | NP_001919.2 | 2 | 154 | ||
GenBank mRNA | AJ313463.1 | - | 2 | 123 | |
AK300963.1 | - | 2 | 149 | ||
BC034529.1 | - | 1 | 70 | ||
BC040146.1 | - | 1 | 56 | ||
BC051001.1 | - | 2 | 94 | ||
BC057807.1 | - | 2 | 153 | 5269709 | |
BG573629.1 | - | 2 | 360 | ||
CR536575.1 | - | 1 | 48 | ||
DA008405.1 | - | 2 | 170 | ||
DQ884409.1 | - | 2 | 144 | ||
HY087483.1 | - | 2 | 155 | ||
M84526.1 | - | 2 | 102 |