Gene Transcripts
- Gene Symbol
- THAP5
- Entrez Gene ID
- 168451
- Gene Name
- THAP domain containing 5
- Gene Aliases
- Chromosome Location
- Chr. 7: 108562144 - 108569768 on Build GRCh38
- UniGene ID
- Hs.707689
- Species
- Homo sapiens
- Gene Symbol
- THAP5P1
- Entrez Gene ID
- 285872
- Gene Name
- THAP domain containing 5 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 158622173 - 158623381 on Build GRCh38
- UniGene ID
- Hs.490790
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126609.1 |
- |
1 |
2006 |
|
View Full Product Details