Gene Transcripts

Gene Symbol
OR2L2
Entrez Gene ID
26246
Gene Name
olfactory receptor family 2 subfamily L member 2
Gene Aliases
HSHTPCRH07, HTPCRH07, OR1-48, OR2L12, OR2L4P
Chromosome Location
Chr. 1: 248035420 - 248039332 on Build GRCh38
UniGene ID
Hs.684888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004686.2 NP_001004686.1 1 331
XM_011544158.2 XP_011542460.1 2 461
GenBank mRNA BC104792.1 - 1 331
BC104794.1 - 1 331 8143797

View Full Product Details