Gene Transcripts
- Gene Symbol
- PCDH11X
- Entrez Gene ID
- 27328
- Gene Name
- protocadherin 11 X-linked
- Gene Aliases
- PCDH-X, PCDH11, PCDHX, PPP1R119
- Chromosome Location
- Chr. X: 91779310 - 92623230 on Build GRCh38
- UniGene ID
- Hs.655673
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168360.1 |
NP_001161832.1
|
2, 3 |
3083 |
|
NM_001168361.1 |
NP_001161833.1
|
2, 3 |
3083 |
|
NM_001168363.1 |
NP_001161835.1
|
2, 3 |
3083 |
|
NM_032968.4 |
NP_116750.1
|
2, 3 |
3083 |
|
NM_032969.4 |
NP_116751.1
|
2, 3 |
3083 |
|
XM_011530910.1 |
XP_011529212.1
|
5, 6 |
3558 |
|
XM_011530911.2 |
XP_011529213.1
|
7, 8 |
3846 |
|
XM_011530912.2 |
XP_011529214.1
|
6, 7 |
3792 |
|
XM_011530913.2 |
XP_011529215.1
|
6, 7 |
3792 |
|
XM_011530914.2 |
XP_011529216.1
|
3, 4 |
3458 |
|
XM_017029416.1 |
XP_016884905.1
|
2, 3 |
3067 |
|
XM_017029417.1 |
XP_016884906.1
|
6, 7 |
3792 |
|
XM_017029418.1 |
XP_016884907.1
|
6, 7 |
3792 |
|
XM_017029421.1 |
XP_016884910.1
|
2, 3 |
3067 |
|
GenBank mRNA |
AB037747.1 |
- |
1, 2 |
271 |
|
AF206516.1 |
- |
2, 3 |
3039 |
|
AF332218.1 |
- |
2, 3 |
3884 |
|
AF332219.1 |
- |
2, 3 |
3884 |
|
AY861432.1 |
- |
2, 3 |
3039 |
|
AY861434.1 |
- |
2, 3 |
3039 |
|
AY861435.1 |
- |
2, 3 |
3039 |
|
View Full Product Details