Gene Transcripts

Gene Symbol
TTC28-AS1
Entrez Gene ID
284900
Gene Name
TTC28 antisense RNA 1
Gene Aliases
TTC28-AS, TTC28AS, dJ353E16.2
Chromosome Location
Chr. 22: 27919376 - 28002679 on Build GRCh38
UniGene ID
Hs.602319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026963.1 3 324
GenBank mRNA AB051435.1 - 2 1141
AL832597.1 - 3 279
CT841515.1 - 2 682

View Full Product Details