Gene Transcripts
- Gene Symbol
- SRRD
- Entrez Gene ID
- 402055
- Gene Name
- SRR1 domain containing
- Gene Aliases
- HC/HCC, SRR1L
- Chromosome Location
- Chr. 22: 26483867 - 26494620 on Build GRCh38
- UniGene ID
- Hs.709914
- Species
- Homo sapiens
- Gene Symbol
- TFIP11
- Entrez Gene ID
- 24144
- Gene Name
- tuftelin interacting protein 11
- Gene Aliases
- NTR1, Spp382, TIP39, bK445C9.6, hNtr1
- Chromosome Location
- Chr. 22: 26491927 - 26512496 on Build GRCh38
- UniGene ID
- Hs.20225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL050258.1 |
- |
15 |
3246 |
|
View Full Product Details