Gene Transcripts

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
Chromosome Location
Chr. 1: 161118072 - 161125446 on Build GRCh38
UniGene ID
Hs.146406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185093.1 NP_001172022.1 6 1094
NM_001185094.1 NP_001172023.1 7 863
NM_005600.2 NP_005591.1 7 862
XM_005245214.2 XP_005245271.1 7 1213
XM_005245215.4 XP_005245272.1 7 874
XM_005245216.3 XP_005245273.1 7 885
GenBank mRNA AF069987.1 - 7 833
AK296606.1 - 6 1058
AK313323.1 - 7 829
BX416745.2 - 7 829
CR541814.1 - 6 757
CR541846.1 - 6 757

View Full Product Details