Gene Transcripts
- Gene Symbol
- NUP54
- Entrez Gene ID
- 53371
- Gene Name
- nucleoporin 54
- Gene Aliases
- Chromosome Location
- Chr. 4: 76114659 - 76148515 on Build GRCh38
- UniGene ID
- Hs.430435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017426.3 |
NP_059122.2
|
3, 4 |
440 |
|
XM_006714237.1 |
XP_006714300.1
|
4, 5 |
417 |
|
XM_011532033.2 |
XP_011530335.1
|
3, 4 |
383 |
|
XM_017008294.1 |
XP_016863783.1
|
3, 4 |
827 |
|
GenBank mRNA |
AF157322.1 |
- |
3, 4 |
321 |
|
AK001517.1 |
- |
3, 4 |
322 |
|
AK222752.1 |
- |
3, 4 |
327 |
|
AK223352.1 |
- |
3, 4 |
323 |
|
AK315160.1 |
- |
3, 4 |
320 |
|
BC012559.1 |
- |
3, 4 |
337 |
|
DA577078.1 |
- |
3, 4 |
440 |
|
View Full Product Details